p-Toluonitril ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Methylbenzoesäurenitril, 4-Methylbenzonitril, p-Methylbenzonitril

B23029.0B B23029.18 B23029.30
B23029.0BEA 341 EUR
B23029.0B B23029.18 B23029.30
p-Toluonitril ≥98%
p-Toluonitril

Purity: 98+% CAS No.: 104-85-8 Molecular Formula: C8H7N Formula Weight: 117.15 Flash Point: 85°(185°F) Odour: Almond-like Boiling Point: 217° Beilstein Reference: 507386 Merck Reference: 14,9538 MDL No.: MFCD00001827

Formel: C₈H₇N
Molecular Weight: 117,15 g/mol
Siedepunkt: 217 °C
Schmelzpunkt: 26…29 °C
Dichte: 0,981
Flammpunkt: 85 °C (185 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001827
CAS-Nummer: 104-85-8
EINECS: 203-244-8
Merck Index: 14,9538

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR