2-(Aminomethyl)pyridin ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(2-Pyridyl)methylamin, 2-Picolamin, 2-Picolylamin, 2-Pyridylmethylamin
B23208.06
B23208.14
B23208.22
B23208.06EA
27.4
EUR
B23208.06
B23208.14
B23208.22
2-(Aminomethyl)pyridin ≥99%
2-(Aminomethyl)pyridine
Purity: 99%
CAS No.: 3731-51-9
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 90°(194°F)
Refractive Index: 1.5450
Odour: Amine-like
pH: 11-12
Melting Point: -20°
Boiling Point: 202-204°
Beilstein Reference: 108054
UN No.: 2735
MDL No.: MFCD00006360
Formel:
C₆H₈N₂ Molecular Weight: 108,14 g/mol Siedepunkt: 202…204 °C Schmelzpunkt: –20 °C Dichte: 1,049 Flammpunkt: 90 °C (194 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006360 CAS-Nummer: 3731-51-9 EINECS: 223-090-5 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...