9-Fluorencarbonsäure ≥97%
Lieferant: Thermo Scientific
Synonyme:
9H-Fluorene-9-carboxylic acid
B23224.06
B23224.14
B23224.22
B23224.06EA
36.7
EUR
B23224.06
B23224.14
B23224.22
9-Fluorencarbonsäure ≥97%
9-Fluorencarbonsäure
Purity: 97%
CAS No.: 1989-33-9
Molecular Formula: C14H10O2
Formula Weight: 210.23
Flash Point: 230°(446°F)
Melting Point: 226-232°
Beilstein Reference: 1911728
MDL No.: MFCD00001136
Formel:
C₁₄H₁₀O₂ Molecular Weight: 210,23 g/mol Siedepunkt: 365 °C (1013 hPa) Schmelzpunkt: 226…232 °C Dichte: 1,333 g/cm³ (20 °C) Flammpunkt: 230 °C (446 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001136 CAS-Nummer: 1989-33-9 EINECS: 217-866-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...