2-Amino-6-methoxybenzothiazol 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
6-Methoxybenzothiazol-2-ylamine
B23380.09
B23380.18
B23380.09EA
35
EUR
B23380.09
B23380.18
2-Amino-6-methoxybenzothiazol 98%
2-Amino-6-methoxybenzothiazole
Purity: 98%
CAS No.: 1747-60-0
Molecular Formula: C8H8N2OS
Formula Weight: 180.23
Odour: Odourless
Melting Point: 161-164°
MDL No.: MFCD00005787
Formel:
C₈H₈N₂OS Molecular Weight: 180,23 g/mol Siedepunkt: 348 °C (1013 hPa) Schmelzpunkt: 161…164 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005787 CAS-Nummer: 1747-60-0 EINECS: 217-130-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...