4-(Trifluormethoxy)-o-toluidin ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Methyl-4-(trifluormethoxy)anilin, 2-Amino-5-(trifluormethoxy)toluol, 4-(Trifluormethoxy)-2-toluidin
B23706.03
B23706.06
B23706.03EA
29.2
EUR
B23706.03
B23706.06
4-(Trifluormethoxy)-o-toluidin ≥97%
4-(Trifluormethoxy)-o-toluidin
Purity: 97+%
CAS No.: 86256-59-9
Molecular Formula: C8H8F3NO
Formula Weight: 191.15
UN No.: 2810
MDL No.: MFCD01631541
Formel:
C₈H₈F₃NO Molecular Weight: 191,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631541 CAS-Nummer: 86256-59-9 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...