N,N'-Diphenyl-p-phenylendiamin 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: DPPD, N,N′-Diphenyl-1,4-phenylenediamine, N,N'-Diphenyl-1,4-phenylenediamine

B23713.06 B23713.14 B23713.22
B23713.06EA 31.9 EUR
B23713.06 B23713.14 B23713.22
N,N'-Diphenyl-p-phenylendiamin 97%
N,N'-Diphenyl-p-phenylendiamin

Purity: 97% CAS No.: 74-31-7 Molecular Formula: C18H16N2 Formula Weight: 260.34 Melting Point: 146-153° Boiling Point: 220-225°/1mm Merck Reference: 14,3329 MDL No.: MFCD00003015

Formel: C₁₈H₁₆N₂
Molecular Weight: 260.34 g/mol
Siedepunkt: 220…225 °C (1 mmHg)
Schmelzpunkt: 146…153 °C
Dichte: 1.28
Lagertemperatur: Raumtemperatur
MDL: MFCD00003015
CAS-Nummer: 74-31-7
EINECS: 200-806-4
Merck Index: 12,03388

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR