Octyltrichlorsilan ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Trichlor(octyl)silan, Octylsilyltrichlorid

B23753.14 B23753.22 B23753.36
B23753.14EA 27.5 EUR
B23753.14 B23753.22 B23753.36
Octyltrichlorsilan ≥97%
Octyltrichlorsilan

Purity: 97% CAS No.: 5283-66-9 Molecular Formula: CH3(CH2)7SiCl3 Formula Weight: 247.67 Flash Point: 96°(204°F) Refractive Index: 1.4470 Odour: Irritating Boiling Point: 225-227° Beilstein Reference: 1744230 UN No.: 1801 MDL No.: MFCD00000488

Formel: C₈H₁₇Cl₃Si
Molecular Weight: 247,67 g/mol
Siedepunkt: 225…227 °C
Dichte: 1,073
Flammpunkt: 96 °C (204 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000488
CAS-Nummer: 5283-66-9
EINECS: 226-112-1
UN: 1801
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR