α-Brom-o-toluonitril ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-(Brommethyl)benzoesäurenitril, o-(Brommethyl)benzonitril, 2-(Brommethyl)benzonitril
B24278.06
B24278.14
B24278.22
B24278.06EA
37.7
EUR
B24278.06
B24278.14
B24278.22
α-Brom-o-toluonitril ≥98%
α-Brom-o-toluonitril
Purity: 98%
CAS No.: 22115-41-9
Molecular Formula: C8H6BrN
Formula Weight: 196.05
Melting Point: 70-74°
Beilstein Reference: 742605
UN No.: 2923
MDL No.: MFCD00001794
Formel:
C₈H₆BrN Molecular Weight: 196.05 g/mol Siedepunkt: 152…155 °C (15 mmHg) Schmelzpunkt: 70…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001794 CAS-Nummer: 22115-41-9 UN: 2923 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...