1,4-Dimethylpiperazin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
sym-Dimethylpiperazine
B24419.14
B24419.22
B24419.14EA
28.5
EUR
B24419.14
B24419.22
1,4-Dimethylpiperazin ≥98%
1,4-Dimethylpiperazin
Purity: 98%
CAS No.: 106-58-1
Molecular Formula: C6H14N2
Formula Weight: 114.19
Flash Point: 21°(69°F)
Refractive Index: 1.4470
Odour: Amine-like
pH: 10
Melting Point: -1°
Boiling Point: 131-132°
Beilstein Reference: 103022
UN No.: 2733
MDL No.: MFCD00006156
Formel:
C₆H₁₄N₂ Molecular Weight: 114,19 g/mol Siedepunkt: 131…132 °C Schmelzpunkt: –1 °C Dichte: 0,844 Flammpunkt: 21 °C (69 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006156 CAS-Nummer: 106-58-1 EINECS: 203-412-0 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...