2,4,6-Triphenylanilin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
5'-Phenyl-[1,1':3',1''-terphenyl]-2'-amine
B24871.03
B24871.06
B24871.14
B24871.03EA
84.3
EUR
B24871.03
B24871.06
B24871.14
2,4,6-Triphenylanilin ≥98%
2,4,6-Triphenylanilin
Purity: 98%
CAS No.: 6864-20-6
Molecular Formula: C24H19N
Formula Weight: 321.43
Melting Point: 122-125°
MDL No.: MFCD00012371
Formel:
C₂₄H₁₉N Molecular Weight: 321,42 g/mol Schmelzpunkt: 122…125 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012371 CAS-Nummer: 6864-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...