3,4-Dimethoxybenzylamin 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Veratrylamin

B25033.06 B25033.14 B25033.22
B25033.06EA 59.3 EUR
B25033.06 B25033.14 B25033.22
3,4-Dimethoxybenzylamin 97%
3,4-Dimethoxybenzylamin

Purity: 97% CAS No.: 5763-61-1 Molecular Formula: C9H13NO2 Formula Weight: 167.21 Flash Point: 110°(230°F) Refractive Index: 1.5560 Boiling Point: 281-284° Beilstein Reference: 511575 UN No.: 2735 MDL No.: MFCD00008116

Formel: C₉H₁₃NO₂
Molecular Weight: 167,21 g/mol
Siedepunkt: 281…284 °C
Dichte: 1,109
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008116
CAS-Nummer: 5763-61-1
EINECS: 227-287-7
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR