2-Brom-5-fluorbenzaldehyd ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
B25071.06
B25071.14
B25071.06EA
60.1
EUR
B25071.06
B25071.14
2-Brom-5-fluorbenzaldehyd ≥98%
2-Bromo-5-fluorobenzaldehyde
Purity: 98%
CAS No.: 94569-84-3
Molecular Formula: C7H4BrFO
Formula Weight: 203.01
Flash Point: 87°(190°F)
Melting Point: 51-56°
MDL No.: MFCD00142872
Formel:
C₇H₄BrFO Molecular Weight: 203,01 g/mol Schmelzpunkt: 51…56 °C Flammpunkt: 87 °C (190 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00142872 CAS-Nummer: 94569-84-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...