1-Brom-5-chlor-2-iodbenzol ≥98%

Lieferant: Thermo Scientific
Achtung

B25409.14 B25409.06
B25409.14EA 162 EUR
B25409.14 B25409.06
1-Brom-5-chlor-2-iodbenzol ≥98%
1-Brom-5-chlor-2-iodbenzol

Purity: 98% CAS No.: 31928-44-6 Molecular Formula: C6H3BrClI Formula Weight: 317.36 MDL No.: MFCD00672944

Formel: C₆H₃BrClI
Molecular Weight: 317,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672944
CAS-Nummer: 31928-44-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR