1-Methylbenzimidazol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Methyl-1H-benzimidazole
B25472.06
B25472.14
B25472.06EA
70.2
EUR
B25472.06
B25472.14
1-Methylbenzimidazol ≥99%
1-Methylbenzimidazole
Purity: 99%
CAS No.: 1632-83-3
Molecular Formula: C8H8N2
Formula Weight: 132.17
Melting Point: 59-62°
Boiling Point: 154°/12mm
MDL No.: MFCD00192275
Formel:
C₈H₈N₂ Molecular Weight: 132.16 g/mol Siedepunkt: 154 °C (12 mmHg) Schmelzpunkt: 59…62 °C Dichte: 1.11 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192275 CAS-Nummer: 1632-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...