Natriumhydrogensulfat, wasserfrei 90+% remainder mainly sodium sulphate

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Natriumbisulfat

B25587.0B B25587.0I B25587.30
B25587.0BEA 60.7 EUR
B25587.0B B25587.0I B25587.30
Natriumhydrogensulfat, wasserfrei 90+% remainder mainly sodium sulphate
Natriumhydrogensulfat

Purity: 90+% CAS No.: 7681-38-1 Molecular Formula: NaHSO4 Formula Weight: 120.06 Odour: Odourless pH: 1.0 Melting Point: 177-180° Merck Reference: 14,8586 UN No.: 3260 MDL No.: MFCD00003531

Formel: HNaO₄S
Molecular Weight: 120.06 g/mol
Schmelzpunkt: 177…180 °C
Dichte: 2.435
Lagertemperatur: Raumtemperatur
MDL: MFCD00003531
CAS-Nummer: 7681-38-1
EINECS: 231-665-7
UN: 3260
ADR: 8 III
Merck Index: 14,8586

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR