2-Brom-4-(trifluormethyl)pyridin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H25748.03 H25748.06
H25748.03EA 111 EUR
H25748.03 H25748.06
2-Brom-4-(trifluormethyl)pyridin ≥97%
2-Bromo-4-(trifluoromethyl)pyridine

Purity: 97% CAS No.: 175205-81-9 Molecular Formula: C6H3BrF3N Formula Weight: 226.00 Flash Point: 73°(164°F) Refractive Index: 1.4780 Boiling Point: 84-85°/14mm MDL No.: MFCD00153085

Formel: C₆H₃BrF₃N
Molecular Weight: 226 g/mol
Siedepunkt: 84…85 °C (14 mmHg)
Dichte: 1.83
Flammpunkt: 73 °C (164 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00153085
CAS-Nummer: 175205-81-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR