6-Fluor-m-toluonitril ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
H26166.03
H26166.06
H26166.03EA
16.1
EUR
H26166.03
H26166.06
6-Fluor-m-toluonitril ≥99%
6-Fluor-m-toluonitril
Purity: 99%
CAS No.: 64113-84-4
Molecular Formula: C8H6FN
Formula Weight: 135.14
Flash Point: 94°(201°F)
Melting Point: 49-52°
UN No.: 3439
MDL No.: MFCD00134542
Formel:
C₈H₆FN Molecular Weight: 135,14 g/mol Schmelzpunkt: 49…52 °C Flammpunkt: 94 °C (201 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134542 CAS-Nummer: 64113-84-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...