4-Pentylanilin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Amylaniline, 4-n-Pentylaniline, 4-Pentylaniline

H26569.06 H26569.14
H26569.06EA 60.1 EUR
H26569.06 H26569.14
4-Pentylanilin ≥98%
4-Pentylanilin

Purity: 98% CAS No.: 33228-44-3 Molecular Formula: C11H17N Formula Weight: 163.26 Flash Point: 110°(230°F) Refractive Index: 1.53 Boiling Point: 130° MDL No.: MFCD00007926

Formel: C₁₁H₁₇N
Molecular Weight: 163.26 g/mol
Siedepunkt: 130 °C
Dichte: 0.919
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007926
CAS-Nummer: 33228-44-3
EINECS: 251-409-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR