2,6-Dimethoxyanilin ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
H27139.03
H27139.06
H27139.03EA
92.3
EUR
H27139.03
H27139.06
2,6-Dimethoxyanilin ≥97%
2,6-Dimethoxyanilin
Purity: 97%
CAS No.: 2734-70-5
Molecular Formula: C8H11NO2
Formula Weight: 153.18
Odour: Characteristic
Melting Point: 76-78°
UN No.: 2811
MDL No.: MFCD00053934
Formel:
C₈H₁₁NO₂ Molecular Weight: 153,18 g/mol Schmelzpunkt: 76…78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053934 CAS-Nummer: 2734-70-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...