1-Hexyl-3-methylimidazoliumchlorid ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
1-n-Hexyl-3-methylimidazolium chloride
H27178.06
H27178.18
H27178.06EA
32.9
EUR
H27178.06
H27178.18
1-Hexyl-3-methylimidazoliumchlorid ≥98%
1-Hexyl-3-methylimidazolium chloride
Purity: 98%
CAS No.: 171058-17-6
Molecular Formula: C10H19N2Cl
Formula Weight: 202.72
Refractive Index: 1.5165
Formel:
C₁₀H₁₉ClN₂ Molecular Weight: 202.73 g/mol Schmelzpunkt: –85 °C Dichte: 1.05 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03093289 CAS-Nummer: 171058-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...