2-Bromisonicotinonitril ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
H27191.03
H27191.06
H27191.03EA
93.4
EUR
H27191.03
H27191.06
2-Bromisonicotinonitril ≥97%
2-Bromisonicotinonitril
Purity: 97%
CAS No.: 10386-27-3
Molecular Formula: C6H3BrN2
Formula Weight: 183.01
Odour: Odourless
Melting Point: 70-74°
MDL No.: MFCD07367879
Formel:
C₆H₃BrN₂ Molecular Weight: 183,01 g/mol Schmelzpunkt: 70…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07367879 CAS-Nummer: 10386-27-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...