1-Pentyl-4-iodbenzol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-(4'-Iodphenyl)pentan, 1-(4-Iodphenyl)pentan, 1-Iod-4-pentylbenzol

H27356.09 H27356.03
H27356.09EA 148 EUR
H27356.09 H27356.03
1-Pentyl-4-iodbenzol ≥98%
1-Pentyl-4-iodbenzol

Purity: 98% CAS No.: 85017-60-3 Molecular Formula: C11H15I Formula Weight: 274.14 Flash Point: 110°(230°F) Boiling Point: 88°/1mm MDL No.: MFCD00079767

Formel: C₁₁H₁₅I
Molecular Weight: 274,14 g/mol
Siedepunkt: 88 °C (1 mmHg)
Dichte: 1,38
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00079767
CAS-Nummer: 85017-60-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR