5-Amino-2-methoxypyridin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

H27528.06 H27528.14
H27528.06EA 38.2 EUR
H27528.06 H27528.14
5-Amino-2-methoxypyridin 98%
5-Amino-2-methoxypyridin

Purity: 98% CAS No.: 6628-77-9 Molecular Formula: C6H8N2O Formula Weight: 124.14 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5735 Melting Point: 29-31° Boiling Point: 85-90°/1mm MDL No.: MFCD00006264

Formel: C₆H₈N₂O
Molecular Weight: 124.14 g/mol
Siedepunkt: 85…90 °C (1 mmHg)
Schmelzpunkt: 29…31 °C
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00006264
CAS-Nummer: 6628-77-9
EINECS: 229-612-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR