(4-(Brommethyl)phenyl)essigsäure ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-(Bromomethyl)phenylacetic acid

H27744.06 H27744.03
H27744.06EA 121 EUR
H27744.06 H27744.03
(4-(Brommethyl)phenyl)essigsäure ≥97%
(4-(Brommethyl)phenyl)essigsäure

Purity: 97% CAS No.: 13737-36-5 Molecular Formula: C9H9BrO2 Formula Weight: 229.07 Melting Point: 178-180° UN No.: 3261 MDL No.: MFCD00010632

Formel: (C₆H₄(CH₂Br))CH₂COOH
Molecular Weight: 229,07 g/mol
Schmelzpunkt: 178…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010632
CAS-Nummer: 13737-36-5
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR