1-Butyl-3-methylimidazoliummethylsulfat ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
[BMIM] [MeSO4], 1-n-Butyl-3-methylimidazolium methyl sulphate, ECOENG 411, BMIMMeSO4
H27754.06
H27754.18
H27754.06EA
36.3
EUR
H27754.06
H27754.18
1-Butyl-3-methylimidazoliummethylsulfat ≥99%
1-Butyl-3-methylimidazolium methyl sulphate
Purity: 99%
CAS No.: 401788-98-5
Molecular Formula: C9H18N2O4S
Formula Weight: 250.32
Refractive Index: 1.4780
Formel:
C₉H₁₈N₂O₄S Molecular Weight: 250,32 g/mol Dichte: 1,21 Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095437 CAS-Nummer: 401788-98-5 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...