Amantadin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Aminotricyclo[3.3.1.1(3,7)]decan, 1-Adamantylamin, 1-Aminoadamantan
H30076.06
H30076.14
H30076.22
H30076.06EA
62.4
EUR
H30076.06
H30076.14
H30076.22
Amantadin 98%
Amantadin
Purity: 98%
CAS No.: 768-94-5
Molecular Formula: C10H17N
Formula Weight: 151.25
Melting Point: ca 200° subl.
MDL No.: MFCD00074732
Formel:
C₁₀H₁₇N Molecular Weight: 151,25 g/mol Siedepunkt: 214 °C (1013 hPa) Schmelzpunkt: ∼ 200 °C (Subl.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00074732 CAS-Nummer: 768-94-5 EINECS: 212-201-2 Merck Index: 12,00389 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...