2-Amino-6-fluorpyridin 97%
Lieferant: Thermo Scientific
|
Gefahr
|
H30629.03
H30629.06
H30629.03EA
77.4
EUR
H30629.03
H30629.06
2-Amino-6-fluorpyridin 97%
2-Amino-6-fluorpyridin
Purity: 97%
CAS No.: 1597-32-6
Molecular Formula: C5H5FN2
Formula Weight: 112.11
Odour: Characteristic
Melting Point: 58-62°
UN No.: 2811
MDL No.: MFCD04114200
Formel:
C₅H₅FN₂ Molecular Weight: 112,11 g/mol Schmelzpunkt: 58…62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114200 CAS-Nummer: 1597-32-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...