4-Fluorphthalsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H31597.03 H31597.09
H31597.03EA 32.5 EUR
H31597.03 H31597.09
4-Fluorphthalsäure ≥98%
4-Fluorphthalsäure

Purity: 98% CAS No.: 320-97-8 Molecular Formula: C8H5FO4 Formula Weight: 184.12 Odour: Odourless Melting Point: 154-158° MDL No.: MFCD06208213

Formel: C₈H₅FO₄
Molecular Weight: 184.12 g/mol
Schmelzpunkt: 154…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06208213
CAS-Nummer: 320-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR