(2,3-Dichlorphenyl)essigsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,3-Dichlorophenylacetic acid
H32763.06
H32763.14
H32763.06EA
25.2
EUR
H32763.06
H32763.14
(2,3-Dichlorphenyl)essigsäure ≥98%
(2,3-Dichlorphenyl)essigsäure
Purity: 98%
CAS No.: 10236-60-9
Molecular Formula: C8H6Cl2O2
Formula Weight: 205.04
Odour: Odourless
Melting Point: 132-136°
MDL No.: MFCD01861393
Formel:
(C₆H₃Cl₂)CH₂COOH Molecular Weight: 205.04 g/mol Schmelzpunkt: 132…136 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861393 CAS-Nummer: 10236-60-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...