3-Nitro-5-(trifluormethyl)anilin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
H32823.03
H32823.09
H32823.03EA
77.7
EUR
H32823.03
H32823.09
3-Nitro-5-(trifluormethyl)anilin ≥98%
3-Nitro-5-(trifluormethyl)anilin
Purity: 98%
CAS No.: 401-94-5
Molecular Formula: C7H5F3N2O2
Formula Weight: 206.13
Melting Point: 77-81°
UN No.: 2811
MDL No.: MFCD00061229
Formel:
C₇H₅F₃N₂O₂ Molecular Weight: 206,12 g/mol Schmelzpunkt: 77…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00061229 CAS-Nummer: 401-94-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...