3-Chlor-1-fluor-4-nitrobenzol ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Chlor-4-fluornitrobenzol

H32906.03 H32906.06 H32906.14
H32906.03EA 37.5 EUR
H32906.03 H32906.06 H32906.14
3-Chlor-1-fluor-4-nitrobenzol ≥98%
3-Chlor-1-fluor-4-nitrobenzol

Purity: 98% CAS No.: 2106-50-5 Molecular Formula: C6H3ClFNO2 Formula Weight: 175.55 Physical Form: Solid Appearance: Yellow Odour: Characteristic Melting Point: 33-37° UN No.: 2811 MDL No.: MFCD03412200

Formel: C₆H₃ClFNO₂
Molecular Weight: 175,55 g/mol
Schmelzpunkt: 33…37 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03412200
CAS-Nummer: 2106-50-5
EINECS: 218-286-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR