2-Chlorthiazol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H50155.03 H50155.06
H50155.03EA 20.3 EUR
H50155.03 H50155.06
2-Chlorthiazol ≥97%
2-Chlorothiazole

Purity: 97% CAS No.: 3034-52-4 Molecular Formula: C3H2ClNS Formula Weight: 119.57 MDL No.: MFCD00210701

Formel: C₃H₂ClNS
Molecular Weight: 119.57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00210701
CAS-Nummer: 3034-52-4
EINECS: 221-228-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR