5-Brom-2-fluor-m-xylol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H50564.03 H50564.06
H50564.03EA 48.2 EUR
H50564.03 H50564.06
5-Brom-2-fluor-m-xylol ≥97%
5-Brom-2-fluor-m-xylol

Purity: 97% CAS No.: 99725-44-7 Molecular Formula: C8H8BrF Formula Weight: 203.06 Boiling Point: 95°/22mm MDL No.: MFCD01320701

Formel: C₈H₈BrF
Molecular Weight: 203.05 g/mol
Siedepunkt: 95 °C (22 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD01320701
CAS-Nummer: 99725-44-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR