tert-Butylcyanacetat ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Cyanessigsäure-tert-butylester
L00123.06
L00123.14
L00123.22
L00123.06EA
42.1
EUR
L00123.06
L00123.14
L00123.22
tert-Butylcyanacetat ≥98%
tert-Butylcyanacetat
Purity: 98%
CAS No.: 1116-98-9
Molecular Formula: NCCH2CO2C(CH3)3
Formula Weight: 141.17
Flash Point: 91°(195°F)
Refractive Index: 1.4200
Odour: Odourless
Boiling Point: 67-68°/1.5mm
Beilstein Reference: 1755933
MDL No.: MFCD00001938
Formel:
C₇H₁₁NO₂ Molecular Weight: 141,17 g/mol Siedepunkt: 67…68 °C (1,5 mmHg) Dichte: 0,987 Flammpunkt: 91 °C (195 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001938 CAS-Nummer: 1116-98-9 EINECS: 214-243-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...