Ethyl-o-toluat ≥98%
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
o-Toluylsäureethylester, 2-Tolylsäureethylester, Ethyl-2-methylbenzoat, 2-Methylbenzoesäureethylester, 2-Toluylsäureethylester
L00598.14
L00598.22
L00598.14EA
37.8
EUR
L00598.14
L00598.22
Ethyl-o-toluat ≥98%
Ethyl-o-toluat
Purity: 98+%
CAS No.: 87-24-1
Molecular Formula: C10H12O2
Formula Weight: 164.21
Flash Point: 91°(195°F)
Refractive Index: 1.5070
Boiling Point: 227°
Beilstein Reference: 2042517
MDL No.: MFCD00009112
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,2 g/mol Siedepunkt: 227 °C Schmelzpunkt: –10 °C Dichte: 1,033 Flammpunkt: 91 °C (195 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009112 CAS-Nummer: 87-24-1 EINECS: 201-734-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...