3-(3-Hydroxyphenyl)propionsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-(3-Hydroxyphenyl)propanoic acid

L01279.06 L01279.14
L01279.06EA 94 EUR
L01279.06 L01279.14
3-(3-Hydroxyphenyl)propionsäure ≥98%
3-(3-Hydroxyphenyl)propionsäure

Purity: 98+% CAS No.: 621-54-5 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 110-114° Beilstein Reference: 1947445

Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Schmelzpunkt: 110…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016554
CAS-Nummer: 621-54-5
EINECS: 210-692-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR