3-Thiophenmethanol ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Thiophen-3-methanol, 3-Thienylmethanol, 3-(Hydroxymethyl)thiophen
L01935.14
L01935.06
L01935.14EA
303
EUR
L01935.14
L01935.06
3-Thiophenmethanol ≥98%
3-Thiophenmethanol
Purity: 98%
CAS No.: 71637-34-8
Molecular Formula: C5H6OS
Formula Weight: 114.17
Flash Point: 110°(230°F)
Refractive Index: 1.5650
Boiling Point: 86-88°/10mm
Beilstein Reference: 106460
MDL No.: MFCD00014534
Formel:
C₅H₆OS Molecular Weight: 114.17 g/mol Siedepunkt: 86…88 °C (10 mmHg) Dichte: 1.210 Flammpunkt: 110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014534 CAS-Nummer: 71637-34-8 EINECS: 275-741-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...