Benzylisocyanat ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
L02633.03
L02633.06
L02633.14
AB174196.0005
L02633.03EA
23.3
EUR
L02633.03
L02633.06
L02633.14
ABCRAB174196.0005
Benzylisocyanat ≥98%
Benzylisocyanat
Purity: 98%
CAS No.: 3173-56-6
Molecular Formula: C8H7NO
Formula Weight: 133.15
Storage Temperature: 2°C to 8°C
Flash Point: 43°(109°F)
Refractive Index: 1.5250
Odour: Pungent
Boiling Point: 53-54°/0.8mm
Beilstein Reference: 636508
UN No.: 3080
MDL No.: MFCD00009701
Formel:
C₈H₇NO Molecular Weight: 133,15 g/mol Siedepunkt: 53…54 °C (0,8 mmHg) Dichte: 1,078 Flammpunkt: 43 °C (109 °F) |
MDL:
MFCD00009701 CAS-Nummer: 3173-56-6 EINECS: 221-640-9 UN: 3080 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...