N-(Brommethyl)phthalimid 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-(Brommethyl)-1,2-benzoldicarbonsäureimid, N-(Brommethyl)phthalsäureimid

L02818.09 L02818.18
L02818.09EA 51.2 EUR
L02818.09 L02818.18
N-(Brommethyl)phthalimid 95%
N-(Brommethyl)phthalimid

Purity: 95% CAS No.: 5332-26-3 Molecular Formula: C9H6BrNO2 Formula Weight: 240.07 Odour: Odourless Melting Point: 144-150° Beilstein Reference: 140943 MDL No.: MFCD00005897

Formel: C₉H₆BrNO₂
Siedepunkt: 410 °C (1013 hPa)
Schmelzpunkt: 144…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005897
CAS-Nummer: 5332-26-3
EINECS: 226-239-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR