6-Aminouracil 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Amino-2,6-dihydroxypyrimidin, 4-Aminouracil, 6-Amino-2,4-dihydroxypyrimidin

L03332.22 L03332.36
L03332.22EA 39.6 EUR
L03332.22 L03332.36
6-Aminouracil 98%
6-Aminouracil

Purity: 98% CAS No.: 873-83-6 Molecular Formula: C4H5N3O2 Formula Weight: 127.10 Odour: Odourless Melting Point: 360° Beilstein Reference: 120491 MDL No.: MFCD00006071

Formel: C₄H₅N₃O₂
Molecular Weight: 127,10 g/mol
Schmelzpunkt: 360 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006071
CAS-Nummer: 873-83-6
EINECS: 212-854-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR