(2-Hydroxyphenoxy)essigsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L03897.06 L03897.14
L03897.06EA 60.5 EUR
L03897.06 L03897.14
(2-Hydroxyphenoxy)essigsäure ≥98%
(2-Hydroxyphenoxy)essigsäure

Purity: 98+% CAS No.: 6324-11-4 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 125-130° Beilstein Reference: 1948566

Formel: (C₆H₄OH)OCH₂COOH
Molecular Weight: 168.15 g/mol
Schmelzpunkt: 125…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015454
CAS-Nummer: 6324-11-4
EINECS: 228-684-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR