N-Ethylisopropylamin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

L04089.03 L04089.06
L04089.03EA 31.3 EUR
L04089.03 L04089.06
N-Ethylisopropylamin 98%
N-Ethylisopropylamin

Purity: 98% CAS No.: 19961-27-4 Molecular Formula: C5H13N Formula Weight: 87.17 Flash Point: 16°(60°F) Refractive Index: 1.3920 Boiling Point: 71-73° Beilstein Reference: 505952 UN No.: 2733 MDL No.: MFCD00015105

Formel: C₅H₁₃N
Molecular Weight: 87.16 g/mol
Siedepunkt: 71…73 °C
Dichte: 0.720
Flammpunkt: 16 °C (60 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00015105
CAS-Nummer: 19961-27-4
EINECS: 243-442-1
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR