5-Amino-2,4(1H,3H)-pyrimidindion 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Aminouracil, 5-Amino-2,4-dihydroxypyrimidin
L04452.06
L04452.14
L04452.06EA
28.8
EUR
L04452.06
L04452.14
5-Amino-2,4(1H,3H)-pyrimidindion 97%
5-Aminouracil
Purity: 97%
CAS No.: 932-52-5
Molecular Formula: C4H5N3O2
Formula Weight: 127.10
Melting Point:
Formel:
C₄H₅N₃O₂ Molecular Weight: 127.1 g/mol Siedepunkt: 377 Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006025 CAS-Nummer: 932-52-5 EINECS: 213-252-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...