Ethylbenzoylacetat ≥90%
Lieferant: Thermo Scientific
Synonyme:
Benzoylessigsäureethylester
L05030.18
L05030.30
L05030.0B
L05030.18EA
39.1
EUR
L05030.18
L05030.30
L05030.0B
Ethylbenzoylacetat ≥90%
Ethylbenzoylacetat
Purity: 90%
CAS No.: 94-02-0
Molecular Formula: C11H12O3
Formula Weight: 192.21
Flash Point: 140°(284°F)
Refractive Index: 1.5290
Boiling Point: 266-270°
Beilstein Reference: 389944
Merck Reference: 14,3767
MDL No.: MFCD00009196
Formel:
C₁₁H₁₂O₃ Molecular Weight: 192,21 g/mol Siedepunkt: 266…270 °C Schmelzpunkt: Max. 0 °C Dichte: 1,113 Flammpunkt: 140 °C (284 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009196 CAS-Nummer: 94-02-0 EINECS: 202-295-3 Merck Index: 14,3767 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...