Methylcholat ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 3α,7α,12α-Trihydroxycholan-24-säuremethylester, Cholsäuremethylester, 3α,7α,12α-Trihydroxycholansäuremethylester, 3α,7α,12α-Trihydroxy-5β-cholansäuremethylester, 3α,7α,12α-Trihydroxy-5β-cholan-24-säuremethylester

L05129.06 L05129.14
L05129.06EA 41.7 EUR
L05129.06 L05129.14
Methylcholat ≥98%
Methylcholat

Purity: 98+% CAS No.: 1448-36-8 Molecular Formula: C25H42O5 Formula Weight: 422.61 Melting Point: 155-156° Beilstein Reference: 2226989 Merck Reference: 14,2203 MDL No.: MFCD00064934

Formel: C₂₅H₄₂O₅
Molecular Weight: 422.61 g/mol
Schmelzpunkt: 155…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064934
CAS-Nummer: 1448-36-8
EINECS: 215-903-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR