N-Ethylbenzylamin 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: N-Benzylethylamin, Benzyl-ethylamin

L05651.AC L05651.AE L05651.AP
L05651.ACEA 39 EUR
L05651.AC L05651.AE L05651.AP
N-Ethylbenzylamin 97%
N-Ethylbenzylamin

Purity: 97% CAS No.: 14321-27-8 Molecular Formula: C6H5CH2NHCH2CH3 Formula Weight: 135.21 Flash Point: 66°(150°F) Refractive Index: 1.5110 Boiling Point: 191-194° Beilstein Reference: 386023 UN No.: 2735 MDL No.: MFCD00009031

Formel: C₆H₅CH₂NHCH₂CH₃
Molecular Weight: 135,21 g/mol
Siedepunkt: 191…194 °C
Dichte: 0,910
Flammpunkt: 66 °C (150 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009031
CAS-Nummer: 14321-27-8
EINECS: 238-265-1
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR