Bis(4-methylphenyl)methanol ≥98%
Lieferant: Thermo Scientific
Synonyme:
4,4'-Dimethyldiphenylmethanol, 4,4'-Dimethylbenzhydrylalkohol, Bis(4-methylphenyl)methanol, 4,4'-Dimethyl-α-phenylbenzylalkohol, Bis(4-methylphenyl)carbinol
L05904.03
L05904.06
L05904.03EA
23.6
EUR
L05904.03
L05904.06
Bis(4-methylphenyl)methanol ≥98%
4,4'-Dimethylbenzhydrol
Purity: 98%
CAS No.: 885-77-8
Molecular Formula: C15H16O
Formula Weight: 212.29
Melting Point: 69-73°
Beilstein Reference: 1874128
Formel:
C₁₅H₁₆O Molecular Weight: 212,29 g/mol Schmelzpunkt: 69…73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017216 CAS-Nummer: 885-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...