1-Phenyl-2-thioharnstoff ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: N-Phenylthioharnstoff

L06690.18 L06690.30 L06690.09
L06690.18EA 109 EUR
L06690.18 L06690.30 L06690.09
1-Phenyl-2-thioharnstoff ≥97%
1-Phenyl-2-thioharnstoff

Purity: 97% CAS No.: 103-85-5 Molecular Formula: C7H8N2S Formula Weight: 152.21 Odour: Odourless Melting Point: 149-153° Beilstein Reference: 907309 Merck Reference: 14,7315 UN No.: 2811 MDL No.: MFCD00004933

Formel: C₇H₈N₂S
Molecular Weight: 152,22 g/mol
Siedepunkt: 295…297 °C (1013 hPa)
Schmelzpunkt: 149…153 °C
Dichte: 1,33 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004933
CAS-Nummer: 103-85-5
EINECS: 203-151-2
UN: 2811
ADR: 6.1,I
Merck Index: 14,7315

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR