2-Amino-1,3-propandiol 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Amino-1,3-dihydroxypropan, Serinol

L08426.06 L08426.14
L08426.06EA 69.4 EUR
L08426.06 L08426.14
2-Amino-1,3-propandiol 98%
2-Amino-1,3-propanediol

Purity: 98% CAS No.: 534-03-2 Molecular Formula: C3H9NO2 Formula Weight: 91.11 Melting Point: 52-56° Boiling Point: 128°/0.8mm UN No.: 3259 MDL No.: MFCD00051487

Formel: C₃H₉NO₂
Molecular Weight: 91.11 g/mol
Siedepunkt: 128 °C (0.8 mmHg)
Schmelzpunkt: 52…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051487
CAS-Nummer: 534-03-2
EINECS: 208-584-0
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR