1-(4-Nitrophenyl)-2-thioharnstoff ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
N-(4-Nitrophenyl)thioharnstoff, 4-Nitrophenylthioharnstoff
L09331.03
L09331.06
L09331.03EA
22.8
EUR
L09331.03
L09331.06
1-(4-Nitrophenyl)-2-thioharnstoff ≥98%
1-(4-Nitrophenyl)-2-thioharnstoff
Purity: 98%
CAS No.: 3696-22-8
Molecular Formula: C7H7N3O2S
Formula Weight: 197.22
Odour: Odourless
Melting Point: ca 207° dec.
Beilstein Reference: 2806574
UN No.: 2811
MDL No.: MFCD00007304
Formel:
(C₆H₄(NO₂))NC(S)NH₂ Molecular Weight: 197,22 g/mol Schmelzpunkt: ∼ 207 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007304 CAS-Nummer: 3696-22-8 EINECS: 223-021-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...